Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1QB, Mouse anti-Human, Janelia Fluor 549, Clone: C1QB/2966, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279922JF549

Catalog No. NB012640


Add to cart

Description

Description

C1QB Monoclonal antibody specifically detects C1QB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

C1QB
C1QB/2966
Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
C1QB
Recombinant fragment (around aa 41-188) of human C1QB protein (exact sequence is proprietary)
Protein A or G purified
RUO
Primary
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit