Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1QL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA563504

Catalog No. PIPA563504


Add to cart

Description

Description

Immunogen sequence: CRMICDPYTA APGGEPPGAK AQPPGPSTAA LEVMQDLSAN PPPPFIQGPK GD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.

Specifications

Specifications

C1QL2
Polyclonal
Unconjugated
C1QL2
Liquid
C1QL2
Recombit protein corresponding to Human C1QL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q7Z5L3
CTRP10
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
165257
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only