Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ C1qR1/CD93 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
$1183.00
Specifications
Assay Range | 15.625 pg/mL |
---|---|
Assay Sensitivity | 15.625-1000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB009839
|
Novus Biologicals™
NBP27940015PLATES |
15 Plates |
Each of 1 for $1,183.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Rat C1qR1/CD93.Specifications
15.625 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Rat | |
C1q receptor 1, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4 | |
CD93 | |
RUO | |
Storage is content dependent. |
15.625-1000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes an antibody specific for Rat C1qR1/CD93. | |
Sandwich ELISA | |
22918 | |
Rabbit | |
B Cell Development and Differentiation Markers | |
C1qR1/CD93 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title