Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1R Mouse anti-Dog, Human, Clone: 1F1, Abnova™

Mouse monoclonal antibody raised against partial recombinant human C1R.

Manufacturer:  Abnova Corporation MAB13010

Catalog No. 89-933-682


Add to cart

Description

Description

Specifications

Specifications

C1R
1F1
Mouse monoclonal antibody raised against partial recombinant human C1R.
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
C1R
Recombinant protein corresponding to amino acids 262-562 of human C1R.
Protein A/G purification
RUO
Primary
715
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
Mouse
IgG2b
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Canine, Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit