Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1S Mouse anti-Human, Clone: OTI2F5, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504081

Catalog No. 50-167-4847


Add to cart

Description

Description

This gene encodes a serine protease, which is a major constituent of the human complement subcomponent C1. C1s associates with two other complement components C1r and C1q in order to yield the first component of the serum complement system. Defects in this gene are the cause of selective C1s deficiency.
Specifications

Specifications

C1S
Monoclonal
0.88 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P09871
EDSPD2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
716
Flow Cytometry, Western Blot
OTI2F5
Unconjugated
C1S
Liquid
C1S
Full length human recombit protein of human C1S produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit