Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1S Mouse anti-Human, Monkey, Mouse, Rat, Clone: 2A8, Abnova™

Mouse monoclonal antibody raised against full length recombinant human C1S.

Manufacturer:  Abnova Corporation MAB13004

Catalog No. 89-933-683


Add to cart

Description

Description

This gene encodes a serine protease, which is a major constituent of the human complement subcomponent C1. C1s associates with two other complement components C1r and C1q in order to yield the first component of the serum complement system. Defects in this gene are the cause of selective C1s deficiency. [provided by RefSeq]

Specifications

Specifications

C1S
2A8
Mouse monoclonal antibody raised against full length recombinant human C1S.
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
FLJ44757
Mouse
IgG2a
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Monkey, Murine, Rat
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
C1S
Recombinant protein corresponding to full length human C1S.
Protein A/G purification
RUO
Primary
716
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit