Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C1S Mouse anti-Human, Mouse, Rat, Clone: OTI2A8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA504247
Description
This gene encodes a serine protease, which is a major constituent of the human complement subcomponent C1. C1s associates with two other complement components C1r and C1q in order to yield the first component of the serum complement system. Defects in this gene are the cause of selective C1s deficiency.Specifications
C1S | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P09871, Q6P6T1 | |
C1S, C1s1 | |
Full length human recombit protein of human C1S produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2A8 | |
Unconjugated | |
C1S | |
EDSPD2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
192262, 50908, 716 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction