Learn More
CERILLIANT™ Colchicine Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco C2071ML
Specifications
75-05-8, 64-86-8 | |
1 mg/mL in acetonitrile | |
Ampule | |
6°C (42.8°F) - closed cup |
C22H25NO6 | |
399.44 | |
1 mL |
Safety and Handling
H332-H319-H340
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation