Learn More
Aviva Systems Biology Corp C20orf10 Antibody - middle region (ARP53699_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53699_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C20orf10. Alternate names: CLG01, TP53TG5, C20orf10ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-61097