Learn More
Aviva Systems Biology Corp C20orf111 Antibody - N-terminal region (ARP56915_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56915_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf111. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C20orf111. Alternate names: HSPC207, Perit1, dJ1183I21.1, C20orf111ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62032