Learn More
Aviva Systems Biology Corp C20orf116 Antibody - N-terminal region (ARP49786_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49786_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf116. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C20orf116. Alternate names: MGC2592, dJ1187M17.3, UFBP1, C20orf116ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit
Catalog No. 50-014-60181