Learn More
Aviva Systems Biology Corp C20orf116 Antibody - middle region (ARP49787_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49787_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf116. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C20orf116. Alternate names: MGC2592, dJ1187M17.3, UFBP1, C20orf116ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-60182