Learn More
C20orf152 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 248701AP
Catalog No. 50-173-4586
Add to cart
Specifications
Specifications
C20orf152
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96M20, Q9D5U8
C20orf152, CNBD2
Rabbit
IgG
150 μL
-20°C
Primary
140894, 70873
Western Blot
Unconjugated
CNBD2
Liquid
Cnbd2
C20orf152 Fusion Protein Ag21668
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse