Learn More
C20orf3 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI4F6, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA504220
Catalog No. 50-167-4947
Add to cart
Description
Description
C20orf3 may play a role in adipocyte differentiation.
Specifications
Specifications
C20orf3
Monoclonal
0.85 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q7TP48, Q9D7N9, Q9HDC9
BSCv, C20orf3
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
366227, 57136, 608089, 71881
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI4F6
Unconjugated
APMAP
Liquid
APMAP
Full length human recombit protein of human C20orf3 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Rat