Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf3 Mouse anti-Human, Clone: OTI1A9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525876

Catalog No. PIMA525876


Add to cart

Description

Description

C20orf3 may play a role in adipocyte differentiation.
Specifications

Specifications

C20orf3
Monoclonal
0.62 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HDC9
C20orf3
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57136
Immunohistochemistry (Paraffin), Western Blot
OTI1A9
Unconjugated
APMAP
Liquid
APMAP
Full length human recombit protein of C20orf3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit