Learn More
C20orf4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 201791AP
Catalog No. 50-173-2750
Add to cart
Description
Description
AAR2 encodes the homolog of the yeast A1-alpha2 repressin protein that is involved in mRNA splicing. Alternately spliced transcript variants have been found for this gene.
Specifications
Specifications
C20orf4
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9D2V5, Q9Y312
C20orf4, CGI 23, PRO0225
Rabbit
IgG
150 μL
-20°C
Primary
25980, 296312, 68295
Western Blot
Unconjugated
AAR2
Liquid
Aar2
C20orf4 Fusion Protein Ag14039
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat