Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATD3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP180552
Description
GATD3A Polyclonal specifically detects GATD3A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
C21orf33 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
chromosome 21 open reading frame 33, D21S2048E, ES1, GT335, HES1, HES1KNP-Ia, human HES1 protein, homolog to E.coli and zebrafish ES1 protein, 10Protein GT335, Keio novel protein I, KNPH, KNPI, KNP-I, KNPImitochondrial, Protein KNP-I | |
Rabbit | |
Protein A purified | |
RUO | |
8209 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
NP_937798 | |
C21ORF33 | |
Synthetic peptide directed towards the N terminal of human C21orf33. Peptide sequence SRGGAEVQIFAPDVPQMHVIDHTKGQPSEGESRNVLTESARIARGKITDL. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Sheep: 92%; Xenopus: 75%. | |
Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Sheep, Zebrafish | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction