Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C21orf59 Mouse anti-Human, Clone: OTI1A1, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501360

Catalog No. 50-167-2503


Add to cart

Description

Description

The function of the C21orf59 protein remains unknown.
Specifications

Specifications

C21orf59
Monoclonal
0.69 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P57076
C21orf48, CILD26, FBB18, Kur
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56683
Immunocytochemistry, Immunofluorescence, Western Blot
OTI1A1
Unconjugated
C21orf59
Liquid
C21orf59
Full length human recombit protein of human C21orf59 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit