Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

C21orf59 Mouse anti-Human, Clone: OTI2H5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525159

Catalog No. PIMA525159



Description

Description

This gene encodes a protein that plays a critical role in dynein arm assembly and motile cilia function. Mutations in this gene result in primary ciliary dyskinesia. Naturally occuring readthrough transcription occurs from this locus to the downstream t-complex 10 like (TCP10L) gene.
Specifications

Specifications

C21orf59
Monoclonal
0.55 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P57076
C21orf59, C21orf48, CILD26, FBB18, Kur
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56683
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2H5
Unconjugated
C21ORF59
Liquid
C21ORF59
Full length human recombit protein of C21orf59 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit