Learn More
C21orf62 Mouse anti-Human, Clone: OTI5B5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808848
Catalog No. 50-166-9121
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
C21orf62 (Chromosome 21 Open Reading Frame 62) is a Protein Coding gene.
Specifications
Specifications
C21orf62
Monoclonal
Unconjugated
C21orf62
Lyophilized
C21orf62
Full length human recombit protein of human C21orf62 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5B5
PBS with 8% trehalose and no preservative; pH 7.3
Q9NYP8
B37, C21orf120, PRED81
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
56245