Learn More
Aviva Systems Biology Corp C21orf66 Antibody - C-terminal region (ARP47636_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47636_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C21orf66. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human C21orf66. Alternate names: BM-020, FLJ90561, GCFC, BM020, FSAP105, C21orf66ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59618