Learn More
Aviva Systems Biology Corp C22orf31 Antibody - N-terminal region (ARP57990_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57990_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C22orf31. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C22orf31. Alternate names: HS747E2A, bK747E2.1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62204