Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2orf47 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549318

Catalog No. PA549318

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

Specifications

Specifications

C2orf47
Polyclonal
PBS with 0.09% sodium azide
Q8WWC4
Uncharacterized protein C2orf47, mitochondrial, C2orf47
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79568
Western Blot
Unconjugated
MAIP1
Liquid
MAIP1
KLH conjugated synthetic peptide between 190-219 amino acids from the C-terminal region of human C2orf47.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit