Learn More
Aviva Systems Biology Corp PRR30 Antibody - middle region (ARP55780_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55780_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C2orf53. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C2orf53. Alternate names: MGC44505ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61608