Learn More
Aviva Systems Biology Corp C2orf60 Antibody - middle region (ARP51218_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51218_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C2orf60. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C2orf60. Alternate names: FLJ37953, MGC70509, hTYW5, C2orf60ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60518