Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C2orf71 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562491
Description
Immunogen sequence: HSDLVNSVAK SGIQFLKKPK AIRPGCQGGS ERGSIPLLVK NSTCYDAGEG LAEEQPSPRR NQTTAKGLCQ LMGDPASGKR KDMEGLIPGT KTSSS Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 42%.
The protein encoded by this gene is highly expressed in photoreceptors and may associate with the primary cilium of the outer segment. The encoded protein appears to undergo post-translational lipid modification. Nonsense and missense variants of this gene appear to cause a recessive form of retinitis pigmentosa.
Specifications
C2orf71 | |
Polyclonal | |
Unconjugated | |
C2orf71 | |
C2orf71; chromosome 2 open reading frame 71; PCARE; Photoreceptor cilium actin regulator; RP54; uncharacterized protein C2orf71 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
388939 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NGG8 | |
C2orf71 | |
Recombinant protein corresponding to Human C2orf71. Recombinant protein control fragment (Product #RP-99718). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction