Tocris™ C34

TLR4 inhibitor

Manufacturer: Tocris™ 5373/10

 View more versions of this product

Catalog No. 53-731-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors C34
    Molecular Weight 389.4
    Quantity 10mg
    Product Type C34

    TLR4 inhibitor; attenuates TLR4 signaling in enterocytes and macrophages in vitro, and reduces systemic inflammation in mouse models of endotoxemia and necrotizing enterocolitis (NEC). Also suppresses LPS signaling in human ileum displaying NEC ex-vivo. Displays no significant effects on signaling via TLR2 or TLR9.

    • Chemical Name: 1-Methylethyl 2-(acetylamino)-2-deoxy-α-D-glucopyranoside 3,4,6-triacetate
    • Formula: C17H27NO9
    • CAS Number: 40592-88-9
    • Solubility: Soluble to 50mM in water and to 100mM in DMSO

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit