Learn More
C3a/C3a des Arg Mouse anti-Human, Clone: 2991, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA533353
Catalog No. PIMA533353
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Application Notes: Western blot - A reduced sample treatment and SDS-PAGE was used. The band size is ∽8 kDa . Positive control: Activated human serum or purified human C3a desArg protein; Negative control: For Western blot, any irrelevant protein can be used.
C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.
Specifications
Specifications
C3a/C3a des Arg
Monoclonal
0.5 mg/mL
PBS with no preservative
P01024
C3, Acylation Stimulating Protein Cleavage Product
Mouse
IgG1
0.5 mg
4° C
Primary
718
ELISA, Western Blot
2991
Unconjugated
C3
liquid
C3
Human C3a des-Arg
Protein G
RUO
Antibody
Monoclonal
Human