Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C3a Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550364
Description
The antibody detects endogenous levels of total C3a Receptor protein.
C3a Receptor Polyclonal antibody specifically detects C3a Receptor in Human samples. It is validated for Western Blot.Specifications
C3a Receptor | |
Polyclonal | |
Unconjugated | |
C3ar1 | |
anaphylatoxin C3a receptor; AZ3B; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; C3a-R; C3AR1; C3r1; complement component 3 receptor 1; complement component 3a receptor 1; HNFAG09 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
719 | |
-20°C | |
Liquid |
Western Blot | |
1.0 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q16581 | |
C3ar1 | |
Fusion protein corresponding to a region derived from internal residues of human complement component 3a receptor 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction