Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3AR1, anti-Human, Clone: 3G7, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00003

Catalog No. 89-032-346


Add to cart

Specifications

Specifications

C3AR1
3G7
ELISA (0.1 ug/mL) Flow Cytometry (1 ug/mL) The optimal working dilution should be determined by the end user.
complement component 3a receptor 1
AZ3B|C3AR|HNFAG09
Original antibody is raised against the 2e loop of human C3AR1.
200 μg
Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
719
Human
ELISA, Flow Cytometry
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
C3AR1
IgG1, kappa
RUO
Antibody
Monoclonal
This antibody binds specifically to the 2e loop of the human C3AR1, residues 185-193.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit