Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3orf15 Mouse anti-Human, Clone: OTI4F2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526540

Catalog No. PIMA526540


Add to cart

Description

Description

May regulate cilium motility through its role in the assembly of the axonemal radial spokes.
Specifications

Specifications

C3orf15
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q7Z4T9
MAATS1; AAT1; C3orf15
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
89876
Immunohistochemistry (Paraffin), Western Blot
OTI4F2
Unconjugated
MAATS1
Liquid
MAATS1
Human recombit protein fragment corresponding to amino acids 1-313 of C3orf15 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit