Learn More
C3orf20 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 209061AP
Catalog No. 50-173-3149
Add to cart
Description
Description
20906-1-AP can recognize both 88 and 101 kDa isoforms.
The function of C3orf20 has not been widely studied, and is yet to be fully elucidated. Two isoforms were produced by alternative splicing with predicted MW of 88 and 101 kDa according to UniProt.
Specifications
Specifications
C3orf20
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8ND61
C3orf20
Rabbit
IgG
150 μL
-20°C
Primary
84077
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
C3orf20
Liquid
C3orf20
C3orf20 Fusion Protein Ag15031
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse