Learn More
C3orf21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 209131AP
Catalog No. 50-173-3152
Add to cart
Description
Description
XXYLT1 gene ontology annotations related to this gene include transferase activity, transferring glycosyl groups.
Specifications
Specifications
C3orf21
0.54 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3U4G3, Q8NBI6
C3orf21, XXYLT1, xyloside xylosyltransferase 1
Rabbit
IgG
150 μL
-20°C
Primary
152002, 268880, 363799
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
XXYLT1
Liquid
Xxylt1
C3orf21 Fusion Protein Ag15043
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat