Learn More
Aviva Systems Biology Corp C3orf24 Antibody - middle region (ARP53229_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53229_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C3orf24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C3orf24. Alternate names: MGC40179, C3orf24ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60979