Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C3orf26 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546096
Description
Peptide sequence: KTKQPKECFL IQPKERKENT TKTRKRRKKK ITDVLAKSEP KPGLPEDLQK Sequence homology: Human: 100%.
CMSS1 (Cms1 Ribosomal Small Subunit Homolog) is a Protein Coding gene. Diseases associated with CMSS1 includeRefractive Error. Gene Ontology (GO) annotations related to this gene includenucleic acid binding.
Specifications
C3orf26 | |
Polyclonal | |
Unconjugated | |
CMSS1 | |
1110001A06Rik; 2610528E23Rik; 4930572F24Rik; AV099812; AV122629; C3orf26; cms small ribosomal subunit 1; cms1 ribosomal small subunit homolog; cms1 ribosomal small subunit homolog (yeast); Cmss1; MGC4308; Protein CMSS1; RGD1309437 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84319 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BQ75 | |
CMSS1 | |
Synthetic peptide directed towards the N-terminal of human C3orf26 (aa 48-97). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction