Learn More
Aviva Systems Biology Corp POMGNT2 Antibody - middle region (ARP48972_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48972_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against C3orf39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C3orf39. Alternate names: AGO61, FLJ14566, C3orf39ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59980