Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C4.4A/LYPD3 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
Supplier: R&D Systems AF5428SP
This item is not returnable.
View return policy
Description
C4.4A/LYPD3 Polyclonal specifically detects C4.4A/LYPD3 in Human samples. It is validated for Western Blot.Specifications
C4.4A/LYPD3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C4.4A2310061G07Rik, GPI-anchored metastasis-associated protein C4.4A homolog, GPI-anchored metastasis-associated protein homolog, LY6/PLAUR domain containing 3, ly6/PLAUR domain-containing protein 3, LYPD3, Matrigel-induced gene C4 protein, MIG-C4 | |
Sheep | |
Affinity Purified | |
RUO | |
27076 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
O95274 | |
LYPD3 | |
Mouse myeloma cell line NS0-derived recombinant human C4.4A/LYPD3 Leu31-His286 Accession # O95274 | |
25 μg | |
Primary | |
Detects human C4.4A/LYPD3 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse C4.4A is observed. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only