Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4 Mouse anti-Human, Alexa Fluor™ 750, Clone: 25300, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP231555AF750

Catalog No. NP231A70

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

C4 Monoclonal antibody specifically detects C4 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

C4
25300
Western Blot, ELISA
Conjugated, Purified
Mouse
IgG1 Kappa
0.1 mL
Primary
Epitope specificity differs from that of NBP2-31554, as determined by inhibition ELISA.
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
C4
C4 isolated from human plasma adsorbed onto aluminum hydroxide gel.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit