Learn More
C4BP Mouse anti-Human, Clone: 10-07, Abnova™
Mouse monoclonal antibody raised against human C4BP.
Manufacturer:  Abnova Corporation MAB12546
Catalog No. 89-933-695
Add to cart
Description
Description
Specifications
Specifications
C4BP
10-07
Mouse monoclonal antibody raised against human C4BP.
In BBS (0.09% sodium azide).
Mouse
IgG2b
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
ELISA, Immunohistochemistry (PFA fixed)
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Native purified human C4BP.
Protein A purification
RUO
Primary
Human