Learn More
C4C Mouse anti-Human, Clone: 10-12, Abnova™
Mouse monoclonal antibody raised against human Complement Component 4c (C4c).
Manufacturer:  Abnova Corporation MAB12759
Catalog No. 89-933-698
Add to cart
Description
Description
Specifications
Specifications
C4C
10-12
Mouse monoclonal antibody raised against human Complement Component 4c (C4c).
In BBS (0.09% sodium azide).
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Flow Cytometry Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
Purified human Complement Component 4c (C4c).
Protein A purification
RUO
Primary
Human