Learn More
C4orf14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 203231AP
Catalog No. 50-173-2827
Add to cart
Description
Description
The protein encoded by this gene is a nuclear-encoded GTPase that functions in the mitochondrion. Upon translation, this protein is imported into the nucleus and then into the nucleolus before being exported to the mitochondrion. The encoded protein is required for oxygen-dependent regulation of mitochondrial respiratory complexes and for mitochondrial protein synthesis.
Specifications
Specifications
C4orf14
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8NC60, Q9JJG9
C4orf14, hAtNOS1, mAtNOS1
Rabbit
IgG
150 μL
-20°C
Primary
289562, 56412, 84273
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NOA1
Liquid
Noa1
C4orf14 Fusion Protein Ag14186
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat