Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C4orf17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546099
Description
Peptide sequence: EIKAKRPPSP PKACSTPGSC SSGMTSTKND VKANTICIPN YLDQEIKILA Sequence homology: Dog: 79%; Human: 100%.
The function of this protein remains unknown.
Specifications
C4orf17 | |
Polyclonal | |
Unconjugated | |
C4orf17 | |
C4orf17; chromosome 4 open reading frame 17; uncharacterized protein C4orf17 | |
Rabbit | |
Affinity chromatography | |
RUO | |
84103 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q53FE4 | |
C4orf17 | |
Synthetic peptide directed towards the middle region of human C4orf17. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction