Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4orf42 Mouse anti-Human, Clone: OTI2C6, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503479

Catalog No. 50-167-4361


Add to cart

Description

Description

C4orf42 (CTBP1-DT, CTBP1 Divergent Transcript) is an RNA Gene, and is affiliated with the non-coding RNA class.
Specifications

Specifications

C4orf42
Monoclonal
4.03 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Liquid
CTBP1-DT
Human recombit protein fragment corresponding to amino acids 1-186 of human C4orf42 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI2C6
Unconjugated
CTBP1-DT
C4orf42, CTBP1-AS1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
92070
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit