Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C4orf46 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548136

Catalog No. PA548136


Add to cart

Description

Description

This gene encodes a small, conserved protein of unknown function that is expressed in a variety of tissues. There are pseudogenes for this gene on chromosomes 6, 8, 16, and X. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Feb 2013]
Specifications

Specifications

C4orf46
Polyclonal
PBS with 0.09% sodium azide
Q3UUX7
Uncharacterized protein C4orf46, C4orf46
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
75939
Western Blot
Unconjugated
4930579G24Rik
Liquid
4930579G24Rik
KLH conjugated synthetic peptide between 84-113 amino acids from the C-terminal region of human C4orf46.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit