Learn More
C5AR1 Mouse anti-Human, Clone: P12/1, Abnova™
Mouse monoclonal antibody raised against human C5AR1.
Manufacturer:  Abnova Corporation MAB12640
Catalog No. 89-933-699
Add to cart
Description
Description
Specifications
Specifications
C5AR1
P12/1
Mouse monoclonal antibody raised against human C5AR1.
In PBS (0.09% sodium azide).
C5A/C5AR/C5R1/CD88
Mouse
IgG2a
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Western Blot The optimal working dilution should be determined by the end user.
Liquid
C5AR1
A synthetic peptide corresponding to human C5AR1.
Protein G purification
RUO
Primary
728