Learn More
C5AR1 Mouse anti-Human, FITC, Clone: P12/1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA528089
 View more versions of this product
Catalog No. PIMA528089
Add to cart
Description
Description
C5AR1 is an C5a Anaphylatoxin Receptor that mediates the effects of one of the most potent chemotactic agents known. C5AR1 has been implicated in asthma, and its expression has been reported in myeloid blood cells, brain, liver, lung, spleen, heart, kidney, and intestinal tract. C5AR1 is the receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. Functionally, C5AR1 stimulates chemotaxis, granule enzyme release and superoxide anion production. C5a is likely a component of the classical pathway of the complement cascade; it may be involved with tissue injury associated with ischemia or sepsis and it may be a component of the membrane attack complex. Diseases associated with C5AR1 include Mannose-Binding Lectin Deficiency and Systemic Lupus Erythematosus.
Specifications
Specifications
C5AR1
Monoclonal
FITC
C5AR1
Liquid
C5AR1
C5aR - peptide Met1- Asn31
Ion-Exchange Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
P12/1
PBS with 1% BSA and 0.09% sodium azide
P21730
CD88
Mouse
IgG2a
25 μg
4° C, store in dark
Primary
728