Learn More
C5orf24 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 243191AP
Catalog No. 50-173-4296
Add to cart
Description
Description
C5orf24 is a protein coding gene.
Specifications
Specifications
C5orf24
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q7Z6I8
C5orf24, UPF0461 protein C5orf24
Rabbit
IgG
150 μL
-20°C
Primary
134553
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
C5orf24
Liquid
C5orf24
C5orf24 Fusion Protein Ag19534
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human