Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5orf33 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548634

Catalog No. PA548634

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, monkey, mouse, pig and rat based on sequence homology.

Mitochondrial NAD(+) kinase that phosphorylates NAD(+) to yield NADP(+). Can use both ATP or inorganic polyphosphate as the phosphoryl donor. Also has weak NADH kinase activity in vitro; however NADH kinase activity is much weaker than the NAD(+) kinase activity and may not be relevant in vivo.
Specifications

Specifications

C5orf33
Polyclonal
PBS with 0.09% sodium azide
Q4G0N4, Q8C5H8
NAD kinase 2, mitochondrial, Mitochondrial NAD kinase, NAD kinase domain-containing protein 1, mitochondrial, NADK2, C5orf33, MNADK, NADKD1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
133686, 68646
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NADK2
Liquid
NADK2
KLH conjugated synthetic peptide between 122-150 amino acids from the Central region of human C5orf33.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit