Learn More
Aviva Systems Biology Corp C5ORF39 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP54435_P050
This is a rabbit polyclonal antibody against C5orf39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C5orf39. Alternate names: AX2R, AXIIR, C5orf39ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53739