Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C5orf54 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565978
Description
Immunogen sequence: ILQQVLEDIKA SPLKVGIQLA ETTDMDDCSQ LMAFVRYIKE REIVEEFLFC EPLQLSMKGI DVFNLFRDFF LKHKIALDVC GSVCTDGASS MLGENSEFV Highest antigen sequence identity to the following orthologs - mouse 45%, rat 48%.
ZBED8 is a protein coding gene.
Specifications
C5orf54 | |
Polyclonal | |
Unconjugated | |
ZBED8 | |
Buster3; C5orf54; Protein ZBED8; transposon-derived Buster3 transposase-like protein; ZBED8; Zinc finger BED domain-containing protein 8; zinc finger BED-type containing 8; zinc finger, BED-type containing 8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
63920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZ13 | |
ZBED8 | |
Recombinant Human C5orf54. Recombinant protein control fragment (Product #RP-106667). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction